Learn More
Trajan Scientific L-MARK Brand Cap Screw Black 8-425 Screw-Thread, PTFE/Silicone, 100 Pack
Catalog No. 501090046
Encompass_Preferred
L-MARK Brand Screw Cap, PP, Black, Red PTFE/White Silicone, 8-425 Chromatography, 100 Pack. Made in the USA.
Catalog No. 50-109-0046 Supplier Trajan Scientific Supplier No. LPP.008138581
L-MARK Brand Screw Cap, PP, Black, Red PTFE/White Silicone, 8-425 Chromatography, 100 Pack. Made in the USA.