Learn More
Trajan Scientific L-MARK Brand Cap Blue Screw 9 mm, 100 Pack
Catalog No. 501090047
Encompass_Preferred
L-MARK Brand Screw Cap, PP, Blue, Red PTFE/White Silicone, 9 mm, Press Fit, 100 Pack. Made in the USA.
Catalog No. 50-109-0047 Supplier Trajan Scientific Supplier No. LPP.009039068
L-MARK Brand Screw Cap, PP, Blue, Red PTFE/White Silicone, 9 mm, Press Fit, 100 Pack. Made in the USA.