Learn More
Trajan Scientific L-MARK Brand Cap Blue Screw Bonded 9 mm, 100 Pack
Catalog No. 501090050
Encompass_Preferred
L-MARK Brand Screw Cap, PP, Blue, Clear PTFE/Clear Silicone LCMS grade, 9 mm, Bonded, 100 Pack. Made in the USA.
Catalog No. 50-109-0050 Supplier Trajan Scientific Supplier No. LPP.009138044
L-MARK Brand Screw Cap, PP, Blue, Clear PTFE/Clear Silicone LCMS grade, 9 mm, Bonded, 100 Pack. Made in the USA.