Learn More
Microsolv Technology Corporation Caps, Screw Top, with Red Butyl Rubber / Grey PTFE Septa, fitted in knurled polypropylene, White Caps. For use with 24-400mm threaded EPA Style vials. ND24 threads. 100/PK.
Catalog No. 502271296
Encompass_Preferred
Septa is 55 Shore A, (Hardness), is 2.5mm thick and can be used as an autosampler vial cap or for long term storage with any 30, 40 or 60 EPA Style vial that has 40-400 threads. Cap has a 12.5mm top hole.
This item is not returnable. View return policy
Catalog No. 50-227-1296 Supplier Microsolv Technology Corporation Supplier No. 9704080C
Septa is 55 Shore A, (Hardness), is 2.5mm thick and can be used as an autosampler vial cap or for long term storage with any 30, 40 or 60 EPA Style vial that has 40-400 threads. Cap has a 12.5mm top hole.