Learn More
Microsolv Technology Corporation Cap Screw Gr Preslit Sil/Ptfe
Catalog No. 5001452173
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52173 Supplier Microsolv Technology Corporation Supplier No. 9502SE35G
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.