Learn More
Microsolv Technology Corporation Cap Screw Green Sil/Ptfe
Catalog No. 502779
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Septa, fitted in knurled, polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 1000/CS.
Catalog No. 50-277-9 Supplier Microsolv Technology Corporation Supplier No. 9502SE15GM
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Septa, fitted in knurled, polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 1000/CS.