Learn More
Trajan Scientific L-MARK Brand Cap Screw 18mm Headspace 1.7mm PTFE/Silicone, 100 Pack
Catalog No. 501090131
Encompass_Preferred
L-MARK Brand Cap Screw 18mm Headspace 1.7mm White PTFE/Trans Blue Silicone Magnetic100 Pack. Made in the USA.
Catalog No. 50-109-0131 Supplier Trajan Scientific Supplier No. LPP.018037715
L-MARK Brand Cap Screw 18mm Headspace 1.7mm White PTFE/Trans Blue Silicone Magnetic100 Pack. Made in the USA.