Learn More
Microsolv Technology Corporation Caps, Screw Top, with PTFE, ultra-pure Septa, fitted in "knurled", polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK
Catalog No. 502270288
Encompass_Preferred
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.
This item is not returnable. View return policy
Catalog No. 50-227-0288 Supplier Microsolv Technology Corporation Supplier No. 9509S50CB
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.