Learn More
Microsolv Technology Corporation Caps, Screw Top, with PTFE Septa, fitted in "knurled", polypropylene, Translucent Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271295
Encompass_Preferred
Rubber-free Cap is useful when rubber cannot be used with your analysis. This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal
This item is not returnable. View return policy
Catalog No. 50-227-1295 Supplier Microsolv Technology Corporation Supplier No. 9502S50CN
Rubber-free Cap is useful when rubber cannot be used with your analysis. This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal