Learn More
Trajan Scientific La-Pha-Pack , Cap Screw Black 13mm 1.0mm Red PTFE/White Silicone/Red PTFE Combination Seal100 Pack. Made in Germany.
Supplier:  Trajan Scientific LAP.13150292
Encompass_Preferred
La-Pha-Pack , Cap Screw Black 13mm 1.0mm Red PTFE/White Silicone/Red PTFE Combination Seal100 Pack. Made in Germany.
Catalog No. 50-711-781