Learn More
Microsolv Technology Corporation Cap Screw Sil/Ptfe Smooth
Catalog No. 5001452069
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52069 Supplier Microsolv Technology Corporation Supplier No. 9502S05B
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.