Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / Polypropylene Septa, in "knurled", Polypropylene Blue Caps. 100/PK.
Catalog No. 502269943
Encompass_Preferred
These unique Screw Caps can be used for almost any sample but are designed for Polyfluoroalkyl Substances (PFAS) testing in our Autosampler Vials. The silicone rubber (for optimal sealing), and Polypropylene fitted Septa (for inertness) are excellent for chromatography. These caps do not contain PTFE like most other autosampler vial caps. Use with AQ™ Brand or RSA™ brand 9mm screw thread autosampler vials for best performance and sealing.
This item is not returnable. View return policy
Catalog No. 50-226-9943 Supplier Microsolv Technology Corporation Supplier No. 9502S10CPS
These unique Screw Caps can be used for almost any sample but are designed for Polyfluoroalkyl Substances (PFAS) testing in our Autosampler Vials. The silicone rubber (for optimal sealing), and Polypropylene fitted Septa (for inertness) are excellent for chromatography. These caps do not contain PTFE like most other autosampler vial caps. Use with AQ™ Brand or RSA™ brand 9mm screw thread autosampler vials for best performance and sealing.