Learn More
Microsolv Technology Corporation Caps, Screw Top, with Tan Silicone Rubber / PTFE Pre-Slit Septa, bonded in "knurled", polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials, 1000/CS.
Catalog No. 502270293
Encompass_Preferred
Pre-slit for easy penetration by autosampler needles and to prevent "Septa Plunge". Recommended for Waters Alliance & Acquity instruments. Ultra-pure Silicone Rubber is a natural Tan color and will not extract pigments. Use this Cap for excellent results in LCMS/MS. Compare to “HPLC Certified” brands.
This item is not returnable. View return policy
Catalog No. 50-227-0293 Supplier Microsolv Technology Corporation Supplier No. 9502S31MBK
Pre-slit for easy penetration by autosampler needles and to prevent "Septa Plunge". Recommended for Waters Alliance & Acquity instruments. Ultra-pure Silicone Rubber is a natural Tan color and will not extract pigments. Use this Cap for excellent results in LCMS/MS. Compare to “HPLC Certified” brands.