Learn More
Microsolv Technology Corporation Caps, Screw Top, with Tan Silicone Rubber / White PTFE Septa, bonded in "knurled", polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270295
Encompass_Preferred
Ultra-pure Septa and is a good choice for LCMS, GCMS & LCMS/MS. this cap is also recommended for Waters Acquity™ autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0295 Supplier Microsolv Technology Corporation Supplier No. 9502S11MG
Ultra-pure Septa and is a good choice for LCMS, GCMS & LCMS/MS. this cap is also recommended for Waters Acquity™ autosamplers.