Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Septa fitted in "knurled", polypropylene, Black Caps. For use with 10-425mm thread, 12x32mm wide mouth autosampler vials. Compare to Shimadzu Caps. 1000/CS
Catalog No. 502270443
Encompass_Preferred
Flange at the bottom of the Cap allows for the vial and cap to be suspended in older Shimadzu autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0443 Supplier Microsolv Technology Corporation Supplier No. 9503SGRM
Flange at the bottom of the Cap allows for the vial and cap to be suspended in older Shimadzu autosamplers.