Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in "knurled", polypropylene, Black Caps. For use with 13-425mm thread, 15x45mm autosampler vials. 100/PK.
Catalog No. 502270445
Encompass_Preferred
Economical Cap with Septa for use in Older Waters instruments that accommodate 4ml autosampler vials.
This item is not returnable. View return policy
Catalog No. 50-227-0445 Supplier Microsolv Technology Corporation Supplier No. 9504S05CBLK
Economical Cap with Septa for use in Older Waters instruments that accommodate 4ml autosampler vials.