Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Septa, bonded in wide mouth, "knurled", polypropylene, Black Caps. For use with 10-425mm threads, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502271317
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Septa, bonded in wide mouth, "knurled", polypropylene, Black Caps. For use with 10-425mm threads, 12x32mm autosampler vials. 1000/CS.
This item is not returnable. View return policy
Catalog No. 50-227-1317 Supplier Microsolv Technology Corporation Supplier No. 9503S10CKM
Caps, Screw Top, with Silicone Rubber / PTFE Septa, bonded in wide mouth, "knurled", polypropylene, Black Caps. For use with 10-425mm threads, 12x32mm autosampler vials. 1000/CS.