Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502271319
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.
Catalog No. 50-227-1319 Supplier Microsolv Technology Corporation Supplier No. 9502S05BM
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.