Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271318
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
This item is not returnable. View return policy
Catalog No. 50-227-1318 Supplier Microsolv Technology Corporation Supplier No. 9502S05K
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in smooth sided, Robo Style, soft polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.