Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in "knurled", polypropylene, Yellow Caps. For use with 9-425 thread, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502271325
Encompass_Preferred
Low durometer makes the Septa easier to penetrate by the autosampler needles.
This item is not returnable. View return policy
Catalog No. 50-227-1325 Supplier Microsolv Technology Corporation Supplier No. 9502S12YM
Low durometer makes the Septa easier to penetrate by the autosampler needles.