Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, fitted in "knurled", polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502271327
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, fitted in "knurled", polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.
This item is not returnable. View return policy
Catalog No. 50-227-1327 Supplier Microsolv Technology Corporation Supplier No. 9502S10CBXM
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, fitted in "knurled", polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.