Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in "knurled", polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271322
Encompass_Preferred
The Septa is bonded to the cap for increased performance of penetration that prevents "septa plunge" and prevents them from falling out or dislodging in the cap during transportation.Very popular and useful for HPLC or GC; save money compared to "Certified" brands.
This item is not returnable. View return policy
Catalog No. 50-227-1322 Supplier Microsolv Technology Corporation Supplier No. 9502S10CG
The Septa is bonded to the cap for increased performance of penetration that prevents "septa plunge" and prevents them from falling out or dislodging in the cap during transportation.Very popular and useful for HPLC or GC; save money compared to "Certified" brands.