Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber /PTFE Pre-Slit Septa, fitted in a smooth sided, Robo Style Yellow Caps. For use with 9-425 thread, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502271329
Encompass_Preferred
Pre-slit for easy penetration by autosampler needles and to prevent "Septa Plunge".
This item is not returnable. View return policy
Catalog No. 50-227-1329 Supplier Microsolv Technology Corporation Supplier No. 9502S32YM
Pre-slit for easy penetration by autosampler needles and to prevent "Septa Plunge".