Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE, ultra-pure Septa, fitted in knurled, polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK
Catalog No. 502270290
Encompass_Preferred
When highest purity in a Septa is required. No color added to the rubber or PTFE (Teflon) for improved purity these caps offer virtually no extractables including colors that can contaminate sensitive analyses. Often chosen for LCMS, GCMS & LCMS/MS. Not recommended for Waters Alliance, Acquity™ and all other autosamplers that require Pre-Slit Septa.
This item is not returnable. View return policy
Catalog No. 50-227-0290 Supplier Microsolv Technology Corporation Supplier No. 9509S10CK
When highest purity in a Septa is required. No color added to the rubber or PTFE (Teflon) for improved purity these caps offer virtually no extractables including colors that can contaminate sensitive analyses. Often chosen for LCMS, GCMS & LCMS/MS. Not recommended for Waters Alliance, Acquity™ and all other autosamplers that require Pre-Slit Septa.