Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, fitted in smooth sided, Robo Style, soft polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271310
Encompass_Preferred
Pre-slit for easy penetration by autosampler needles and to prevent "Septa Plunge" into vials.
This item is not returnable. View return policy
Catalog No. 50-227-1310 Supplier Microsolv Technology Corporation Supplier No. 9502S35K
Pre-slit for easy penetration by autosampler needles and to prevent "Septa Plunge" into vials.