Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit, 40mil Septa, bonded in wide mouth, "knurled", polypropylene, Black Caps. For use with 10-425mm threads, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502271316
Encompass_Preferred
Pre-slit for easy penetration by autosampler needles. This Septa is slightly thinner than most Septa to enhance the ease or needle penetration. 40mil equals 1.016mm.
This item is not returnable. View return policy
Catalog No. 50-227-1316 Supplier Microsolv Technology Corporation Supplier No. 9503S35CKM
Pre-slit for easy penetration by autosampler needles. This Septa is slightly thinner than most Septa to enhance the ease or needle penetration. 40mil equals 1.016mm.