Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in "knurled", polypropylene, White Caps. For use with 13-425mm thread, 15x45mm autosampler vials. 100/PK.
Catalog No. 502270439
Encompass_Preferred
Pre-slit for easy penetration by autosampler needles. Recommended for older Waters instruments. Use with 15x45mm, 4ml vials.
This item is not returnable. View return policy
Catalog No. 50-227-0439 Supplier Microsolv Technology Corporation Supplier No. 9504S30CW
Pre-slit for easy penetration by autosampler needles. Recommended for older Waters instruments. Use with 15x45mm, 4ml vials.