Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, fitted in "knurled", polypropylene, Black Caps. For use with 8-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270442
Encompass_Preferred
Wider hole for use with narrow mouth, 2ml vials. Wider hole provides a larger target area for the autosampler needle during injection.
This item is not returnable. View return policy
Catalog No. 50-227-0442 Supplier Microsolv Technology Corporation Supplier No. 9582S03B
Wider hole for use with narrow mouth, 2ml vials. Wider hole provides a larger target area for the autosampler needle during injection.