Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in "knurled", polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS
Catalog No. 502271306
Encompass_Preferred
Septa bonded to cap for increased performance of penetration and to prevent "Septa Plunge" into vials during injection. Designed for Waters Alliance & Acquity instruments.
This item is not returnable. View return policy
Catalog No. 50-227-1306 Supplier Microsolv Technology Corporation Supplier No. 9502S30CBLM
Septa bonded to cap for increased performance of penetration and to prevent "Septa Plunge" into vials during injection. Designed for Waters Alliance & Acquity instruments.