Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in wide mouth, "knurled", polypropylene, Black Caps. For use with 10-425mm threads, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502271309
Encompass_Preferred
Pre-slit for easy penetration by autosampler needles.
This item is not returnable. View return policy
Catalog No. 50-227-1309 Supplier Microsolv Technology Corporation Supplier No. 9503S30CKM
Pre-slit for easy penetration by autosampler needles.