Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE, ultra-pure, Pre-Slit Septa, fitted in "knurled", polypropylene, Light Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502270291
Encompass_Preferred
Highest purity Septa on the market, with no color added to the Rubber or Teflon for improved purity. Good choice for LCMS & LCMS/MS.Pre-slit for easy penetration by autosampler needles and recommended for Waters Alliance, Acquity™ and all other autosamplers that require Pre-Slit Septa.
This item is not returnable. View return policy
Catalog No. 50-227-0291 Supplier Microsolv Technology Corporation Supplier No. 9509S30CBM
Highest purity Septa on the market, with no color added to the Rubber or Teflon for improved purity. Good choice for LCMS & LCMS/MS.Pre-slit for easy penetration by autosampler needles and recommended for Waters Alliance, Acquity™ and all other autosamplers that require Pre-Slit Septa.