Learn More
Microsolv Technology Corporation Caps, Screw Top, with Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit Septa, bonded in "knurled", polypropylene, Purple Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK
Catalog No. 502270460
Encompass_Preferred
This is our Pre-Slit Screw Cap for autosampler vials. Pre-slit for easy penetration by autosampler needles. Recommended for Waters Alliance & Acquity instruments. Soft-Guard™ Septa are easier to penetrate by autosampler needles, and re-seals better than other Caps on the market. This provides longer instrument up-time, fewer missed injections and more consistent data. Use for HPLC, GC, LCMS or GCMS.
This item is not returnable. View return policy
Catalog No. 50-227-0460 Supplier Microsolv Technology Corporation Supplier No. 9502SE30CP
This is our Pre-Slit Screw Cap for autosampler vials. Pre-slit for easy penetration by autosampler needles. Recommended for Waters Alliance & Acquity instruments. Soft-Guard™ Septa are easier to penetrate by autosampler needles, and re-seals better than other Caps on the market. This provides longer instrument up-time, fewer missed injections and more consistent data. Use for HPLC, GC, LCMS or GCMS.