Learn More
Microsolv Technology Corporation Caps, Screw Top, with Soft-Guard, White Silicone Rubber / Red PTFE Septa, bonded in "knurled", polypropylene, Natural Translucent Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK
Catalog No. 502270472
Encompass_Preferred
Soft-Guard™ Septa are easier to penetrate by autosampler needles, and re-seals better than other Caps on the market. This provides longer instrument up-time, fewer missed injections and more consistent data. Use for HPLC, GC, LCMS or GCMS. Click HERE for more information about Soft-Guard™ Septa Technology.
Catalog No. 50-227-0472 Supplier Microsolv Technology Corporation Supplier No. 9502SE10CN
Soft-Guard™ Septa are easier to penetrate by autosampler needles, and re-seals better than other Caps on the market. This provides longer instrument up-time, fewer missed injections and more consistent data. Use for HPLC, GC, LCMS or GCMS. Click HERE for more information about Soft-Guard™ Septa Technology.