Learn More
Microsolv Technology Corporation Caps, Screw Top, with Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape) Septa, fitted in "knurled", polypropylene, Purple Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK
Catalog No. 502270451
Encompass_Preferred
Screw Thread: 9-425mmMaterial: polypropylene, cap, silicone/PTFE septaFeatures: Cross pre-slit, bonded septa Soft-Guard cap for easier penetration of autosampler needles and resealing. Cross slit allows needles to hit the strike target area. Bonded septa prevents septa from falling out of the cap or into the vial
This item is not returnable. View return policy
Catalog No. 50-227-0451 Supplier Microsolv Technology Corporation Supplier No. 9502SE3XP
Screw Thread: 9-425mmMaterial: polypropylene, cap, silicone/PTFE septaFeatures: Cross pre-slit, bonded septa Soft-Guard cap for easier penetration of autosampler needles and resealing. Cross slit allows needles to hit the strike target area. Bonded septa prevents septa from falling out of the cap or into the vial