Learn More
Microsolv Technology Corporation Cap Screw Top Blue Closed Top
Catalog No. 5001452087
Encompass_Preferred
Caps, Screw Top, Solid, with an ultra-clean, Silicone Rubber / PTFE liner, in knurled, closed Blue Caps. For use with 9-425mm thread,12x32mm autosampler vials. MicroSolv Brand 100/PK.
Catalog No. 50-014-52087 Supplier Microsolv Technology Corporation Supplier No. 9502S09SP
Caps, Screw Top, Solid, with an ultra-clean, Silicone Rubber / PTFE liner, in knurled, closed Blue Caps. For use with 9-425mm thread,12x32mm autosampler vials. MicroSolv Brand 100/PK.