Learn More
CELLTREAT Scientific Products CAP ONLY, Screw Top Micro Tube Cap, Grip Cap With Integrated O-Ring, Green, Non-sterile, 1000/CS
SDP
Catalog No. 501535114
Encompass_Preferred
CELLTREAT Scientific Products Screw Top Micro Tube Grip Cap, Green. Premium Grip Cap features molded TPE sealing rings which ensure O-ring will never slip out of place; striped TPE grips on cap edge enable secure handling under all conditions. Caps are available in various colors for easy identification and color-coding. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Easy 3/4 turn to open and close. Temperature range of -196¿C to 121¿C (not for immersion in liquid nitrogen). Grip caps manufactured from polypropylene with an inert thermoplastic elastomer (TPE) sealing ring and grip ribs. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/re-sealable bag, 1,000/case.
Catalog No. 50-153-5114 Supplier CELLTREAT Scientific Products Supplier No. 230840G
CELLTREAT Scientific Products Screw Top Micro Tube Grip Cap, Green. Premium Grip Cap features molded TPE sealing rings which ensure O-ring will never slip out of place; striped TPE grips on cap edge enable secure handling under all conditions. Caps are available in various colors for easy identification and color-coding. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Easy 3/4 turn to open and close. Temperature range of -196¿C to 121¿C (not for immersion in liquid nitrogen). Grip caps manufactured from polypropylene with an inert thermoplastic elastomer (TPE) sealing ring and grip ribs. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/re-sealable bag, 1,000/case.