Learn More
Microsolv Technology Corporation Cap Screw Yel Preslit Sil/Ptfe
Catalog No. 5001452179
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Yellow Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52179 Supplier Microsolv Technology Corporation Supplier No. 9502SE35Y
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Yellow Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.