Learn More
CELLTREAT Scientific Products CAP ONLY, Screw Top Micro Tube Cap, O-Ring, Translucent, Clear, Non-sterile, 1000/CS
SDP
Catalog No. 501535128
Encompass_Preferred
Natural color Translucent Cap O-rings ensure leak-proof seal, and are a highly visible contrasting color. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Packaged sterile in re-sealable bags.
Catalog No. 50-153-5128 Supplier CELLTREAT Scientific Products Supplier No. 230842C
Natural color Translucent Cap O-rings ensure leak-proof seal, and are a highly visible contrasting color. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Packaged sterile in re-sealable bags.