Learn More
CELLTREAT Scientific Products CAP ONLY, Screw Top Micro Tube Cap, O-Ring, Translucent, Orange, Non-sterile, 1000/CS
SDP
Catalog No. 501535130
Encompass_Preferred
CELLTREAT Scientific Products Screw Top Micro Tube Cap with O-Ring, Translucent Orange. Translucent Cap O-rings ensure leak-proof seal, and are a highly visible contrasting color. Caps are available in various colors for easy identification and color-coding. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Easy 3/4 turn to open and close. Temperature range of -196¿C to 121¿C (not for immersion in liquid nitrogen). Manufactured from polypropylene with synthetic rubber o-ring. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/re-sealable bag, 1,000/case.
Catalog No. 50-153-5130 Supplier CELLTREAT Scientific Products Supplier No. 230842N
CELLTREAT Scientific Products Screw Top Micro Tube Cap with O-Ring, Translucent Orange. Translucent Cap O-rings ensure leak-proof seal, and are a highly visible contrasting color. Caps are available in various colors for easy identification and color-coding. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Easy 3/4 turn to open and close. Temperature range of -196¿C to 121¿C (not for immersion in liquid nitrogen). Manufactured from polypropylene with synthetic rubber o-ring. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 500/re-sealable bag, 1,000/case.