Learn More
Waters Corp Waters Snap Neck Cap, Black, 12 × 32 mm, PTFE/Silicone
Catalog No. 50781265 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product consists of a black snap neck cap and a PTFE/silicone septum, designed for various vial sizes, ideal for sample preparation.
  • Snap neck closure
  • Black cap color
  • 12 × 32 mm dimensions
  • PTFE/silicone seal material
  • Includes cap and septum
  • Suitable for sample preparation
  • 100 units per package
Catalog No. 50-781-265 Supplier Waters Corp Supplier No. 186002649
This product consists of a black snap neck cap and a PTFE/silicone septum, designed for various vial sizes, ideal for sample preparation.
  • Snap neck closure
  • Black cap color
  • 12 × 32 mm dimensions
  • PTFE/silicone seal material
  • Includes cap and septum
  • Suitable for sample preparation
  • 100 units per package