Learn More
Waters Corp Waters Cap and Septum, Screw Neck, Black, PTFE/Silicone, 100-Pack
Catalog No. 50819088 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Waters cap and septum assembly is designed for various vial sizes and offers reliable performance for sample preparation. It features a black screw neck cap and a PTFE/silicone septum, supplied in a convenient pack of 100.
  • Screw neck closure type.
  • Black cap color.
  • PTFE/silicone seal material.
  • Includes both cap and septum.
  • Suitable for sample preparation applications.
  • Each package contains 100 units.
  • Compatible with a wide range of solvents, including common organic solvents, acids, and bases.
  • Designed for compatibility with both manual and automated sample handling processes.
  • Recommended storage temperature range is -40°C to 120°C.
  • Not supplied sterile; sterilization is required before use if needed.
  • Not recommended for reuse after initial use to ensure sample integrity.
Catalog No. 50-819-088 Supplier Waters Corp Supplier No. WAT094174
This Waters cap and septum assembly is designed for various vial sizes and offers reliable performance for sample preparation. It features a black screw neck cap and a PTFE/silicone septum, supplied in a convenient pack of 100.
  • Screw neck closure type.
  • Black cap color.
  • PTFE/silicone seal material.
  • Includes both cap and septum.
  • Suitable for sample preparation applications.
  • Each package contains 100 units.
  • Compatible with a wide range of solvents, including common organic solvents, acids, and bases.
  • Designed for compatibility with both manual and automated sample handling processes.
  • Recommended storage temperature range is -40°C to 120°C.
  • Not supplied sterile; sterilization is required before use if needed.
  • Not recommended for reuse after initial use to ensure sample integrity.