Learn More
Trajan Scientific L-MARK Brand Cap Snap Natural 11 mm Pre-slit, 100 Pack
Catalog No. 501090061
Encompass_Preferred
L-MARK Brand Snap Cap, PE, Natural, 11 mm, Pre-slit Red PTFE/Silicone, Chromatography, 100 Pack. Made in the USA.
Catalog No. 50-109-0061 Supplier Trajan Scientific Supplier No. LPP.011038303
L-MARK Brand Snap Cap, PE, Natural, 11 mm, Pre-slit Red PTFE/Silicone, Chromatography, 100 Pack. Made in the USA.