Learn More
Microsolv Technology Corporation Caps, Snap Top, with Soft-Guard, White Silicone Rubber / Red PTFE Septa, fitted in smooth, polyethylene, Black Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502270381
Encompass_Preferred
Soft-Guard™ Septa provide easier penetration of autosampler needles and better resealing than other brands, for longer instrument up-time and fewer missed injections. This is our Snap Cap.
This item is not returnable. View return policy
Catalog No. 50-227-0381 Supplier Microsolv Technology Corporation Supplier No. 9502CE10K
Soft-Guard™ Septa provide easier penetration of autosampler needles and better resealing than other brands, for longer instrument up-time and fewer missed injections. This is our Snap Cap.