Learn More
Microsolv Technology Corporation Caps, Snap Top, with Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape) Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Catalog No. 502270412
Encompass_Preferred
Soft-Guard™ Septa provide easier penetration of autosampler needles and better resealing than other brands, for longer instrument up-time and fewer missed injections. Pre-Slit Septa are normally recommended for Waters Alliance and Acquity instruments. X Shape Pre-Slit provides a greater target area for needles and are easier to penetrate but do not reseal as well as single slit septa.
This item is not returnable. View return policy
Catalog No. 50-227-0412 Supplier Microsolv Technology Corporation Supplier No. 9502CE30KXM
Soft-Guard™ Septa provide easier penetration of autosampler needles and better resealing than other brands, for longer instrument up-time and fewer missed injections. Pre-Slit Septa are normally recommended for Waters Alliance and Acquity instruments. X Shape Pre-Slit provides a greater target area for needles and are easier to penetrate but do not reseal as well as single slit septa.