Learn More
Microsolv Technology Corporation Caps, Snap Top, with Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape) Septa, fitted in smooth, polyethylene, Red Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Supplier:  Microsolv Technology Corporation 9502CE30RXM
Encompass_Preferred
Soft-Guard™ Septa provide easier penetration of autosampler needles and better resealing than other brands, for longer instrument up-time and fewer missed injections. Pre-Slit Septa are normally recommended for Waters Alliance and Acquity instruments. X Shape Pre-Slit provides a greater target area for needles and are easier to penetrate but do not reseal as well as single slit septa.
Catalog No. 50-227-0418