Learn More
Microsolv Technology Corporation Caps, Snap Top, with PTFE, ultra-pure, 2mil Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm, Snap Top autosampler vials. 100/PK.
Catalog No. 502270303
Encompass_Preferred
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.
This item is not returnable. View return policy
Catalog No. 50-227-0303 Supplier Microsolv Technology Corporation Supplier No. 9509C50CB
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal.