Learn More
Microsolv Technology Corporation Cap Snap Top Sil/Ptfe Blue. Ba
Catalog No. 5001452002
Encompass_Preferred
Caps, Snap Top, with 0.04"" thick, White Silicone Rubber / Natural PTFE Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52002 Supplier Microsolv Technology Corporation Supplier No. 9502CE15B
Caps, Snap Top, with 0.04"" thick, White Silicone Rubber / Natural PTFE Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. BASIK Brand. 100/PK.