Learn More
Microsolv Technology Corporation Cap Snap Top Single Slit Sil/P
Catalog No. 5001452020
Encompass_Preferred
Caps, Snap Top, with 0.04"" thick, White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52020 Supplier Microsolv Technology Corporation Supplier No. 9502CE35G
Caps, Snap Top, with 0.04"" thick, White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. BASIK Brand. 100/PK.