Learn More
Chemglass Life Sciences Cap, Solid, 13-425, Green, TFE Septa
SDP
Catalog No. 50143882
Encompass_Preferred
Cap, Solid, 13-425, Green, TFE Septa. Replacement caps for 1 and 2 dram vials.
Catalog No. 50-143-882 Supplier Chemglass Life Sciences Supplier No. CG491110
Cap, Solid, 13-425, Green, TFE Septa. Replacement caps for 1 and 2 dram vials.