Learn More
Chemglass Life Sciences Cap, Solid, 15-425, Green, TFE Septa
SDP
Catalog No. 50143883
Encompass_Preferred
Cap, Solid, 15-425, Green, TFE Septa. Replacement caps for 1 and 2 dram vials.
Catalog No. 50-143-883 Supplier Chemglass Life Sciences Supplier No. CG491112
Cap, Solid, 15-425, Green, TFE Septa. Replacement caps for 1 and 2 dram vials.